accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104113-71-5

IUPAC Name: 1-(naphthalen-1-yl)piperazine hydrochloride
Synonyms: naphthylpiperazine hydrochloride
Molecular Weight (g/mol): 248.75
Molecular Formula: C14H17ClN2
InChi Key: ZYVYPNZFOCZLEM-UHFFFAOYSA-N
SMILES: Cl.C1CN(CCN1)C1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results