Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(1-Naphthyl)piperazine hydrochloride, Thermo Scientific Chemicals

Catalog No. AAJ6209706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6209706 5 g
AAJ6209703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6209706 Supplier Thermo Scientific Chemicals Supplier No. J6209706
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 104113-71-5 | Molecular Formula: C14H17ClN2 | Molecular Weight: 248.754 g/mol

5-HT1 serotonin receptor agonist. 5-HT2 serotonin receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • 5-HT1 serotonin receptor agonist
  • 5-HT2 serotonin receptor antagonist
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104113-71-5
Molecular Formula C14H17ClN2
Molecular Weight (g/mol) 248.754
MDL Number MFCD00055161
InChI Key ZYVYPNZFOCZLEM-UHFFFAOYSA-N
Synonym 1-1-naphthyl piperazine hydrochloride, 1-naphthalen-1-yl piperazine hydrochloride, naphthylpiperazine hydrochloride, 1-naphthalen-1-ylpiperazine hydrochloride, 1-naphthalen-1-yl-piperazine hydrochloride, piperazine,1-1-naphthalenyl, zlchem 1062, 1-1-naphthyl piperazine hcl, 1-naphthalen-1-yl-piperazine hcl salt, n-1-naphthyl piperazine hydrochloride
PubChem CID 2737391
IUPAC Name 1-naphthalen-1-ylpiperazine;hydrochloride
SMILES C1CN(CCN1)C2=CC=CC3=CC=CC=C32.Cl

Specifications

Color Purple
Quantity 5 g
Formula Weight 248.75
Physical Form Solid
Chemical Name or Material 1-(1-Naphthyl)piperazine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.