CAS RN 1041438-68-9
CAS RN 1041438-68-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ STAT3 Inhibitor VII
The STAT3 Inhibitor VII, also referenced under CAS 1041438-68-9, controls the biological activity of STAT3.