Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ STAT3 Inhibitor VII

Catalog No. 5731035MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-310-35MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-310-35MG Supplier MilliporeSigma™ Supplier No. 5731035MG
Add to Cart
Add to Cart

The STAT3 Inhibitor VII, also referenced under CAS 1041438-68-9, controls the biological activity of STAT3.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Light Orange
Description 95% by HPLC
CAS 1041438-68-9
Molecular Formula C19H7F7N2O3
Formula Weight 444.3g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.