accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CONH3CCH3H3COCH3(Z)(Z)

CAS RN 104162-48-3

IUPAC Name: {2,3-bis[(9Z)-octadec-9-en-1-yloxy]propyl}trimethylazanium chloride
Synonyms: {2,3-bis[(9Z)-octadec-9-en-1-yloxy]propyl}trimethylazanium chloride
Molecular Weight (g/mol): 670.59
Molecular Formula: C42H84ClNO2
InChi Key: LDGWQMRUWMSZIU-LQDDAWAPNA-M
SMILES: [Cl-].CCCCCCCC\C=C/CCCCCCCCOCC(C[N+](C)(C)C)OCCCCCCCC\C=C/CCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results