Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl[2,3-(dioleyloxy)propyl]ammonium Chloride 90.0+%, TCI America™
SDP

Catalog No. T13071G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T13071G Supplier TCI America Supplier No. T13071G

missing translation for 'validMainframeMsg'

Specifications

CAS 104162-48-3
Molecular Formula C42H84ClNO2
Quantity 1 g
MDL Number MFCD00145456
Synonym N-[1-(2,3-Dioleyloxy)propyl]-N,N,N-trimethylammonium Chloride, DOTMA
InChI Key LDGWQMRUWMSZIU-LQDDAWAPNA-M
SMILES [Cl-].CCCCCCCC\C=C/CCCCCCCCOCC(C[N+](C)(C)C)OCCCCCCCC\C=C/CCCCCCCC
IUPAC Name {2,3-bis[(9Z)-octadec-9-en-1-yloxy]propyl}trimethylazanium chloride
Molecular Weight (g/mol) 670.59
PubChem CID 16730297
Formula Weight 670.59
Percent Purity ≥90.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material Trimethyl[2,3-(dioleyloxy)propyl]ammonium Chloride
Show More Show Less
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.