accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104197-14-0

IUPAC Name: 5-bromo-1,3-difluoro-2-methoxybenzene
Synonyms: 5-bromo-1,3-difluoro-2-methoxybenzene
Molecular Weight (g/mol): 223.02
Molecular Formula: C7H5BrF2O
InChi Key: CDOQKISJPOWBKC-UHFFFAOYSA-N
SMILES: COC1=C(F)C=C(Br)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results