Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2,6-difluoroanisole 98.0+%, TCI America™
SDP

Catalog No. B33915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B33915G Supplier TCI America Supplier No. B33915G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 104197-14-0
Molecular Formula C7H5BrF2O
Molecular Weight (g/mol) 223.02
MDL Number MFCD01631351
InChI Key CDOQKISJPOWBKC-UHFFFAOYSA-N
Synonym 4-bromo-2,6-difluoroanisole, 5-bromo-1,3-difluoro-2-methoxy-benzene, 4-bromo-2,6-difluorophenyl methyl ether, benzene, 5-bromo-1,3-difluoro-2-methoxy, pubchem8361, acmc-1bo3j, 4-bromo-2,6-difluoranisole, ksc183c9b, 4-bromo-2,6-difluoro-anisol, 3,5-difluoro-4-methoxy-bromobenzene
PubChem CID 2773287
IUPAC Name 5-bromo-1,3-difluoro-2-methoxybenzene
SMILES COC1=C(F)C=C(Br)C=C1F

Specifications

Boiling Point 100°C
Quantity 5 g
Formula Weight 223.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-2,6-difluoroanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.