accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONNNNH2NOCH3O

CAS RN 104227-87-4

IUPAC Name: 2-[(acetyloxy)methyl]-4-(2-amino-9H-purin-9-yl)butyl acetate
Synonyms: famciclovir famvir 泛昔洛韦 famciclovirum FCV
Molecular Weight (g/mol): 321.34
Molecular Formula: C14H19N5O4
InChi Key: GGXKWVWZWMLJEH-UHFFFAOYSA-N
SMILES: CC(=O)OCC(CCN1C=NC2=CN=C(N)N=C12)COC(C)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)