Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Famciclovir

Catalog No. AC463170050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463170050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC463170050 Supplier Thermo Scientific Chemicals Supplier No. 463170050

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104227-87-4
Molecular Formula C14H19N5O4
Molecular Weight (g/mol) 321.34
MDL Number MFCD00866964
InChI Key GGXKWVWZWMLJEH-UHFFFAOYSA-N
IUPAC Name 2-[(acetyloxy)methyl]-4-(2-amino-9H-purin-9-yl)butyl acetate
SMILES CC(=O)OCC(CCN1C=NC2=CN=C(N)N=C12)COC(C)=O

Specifications

Alpha Vector FAMCICLOVIR
Melting Point 103°C to 107°C
Color White
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 5 g
Sensitivity Heat sensitive
Formula Weight 321.33
Physical Form Crystalline Powder
Chemical Name or Material Famciclovir
ShelfLife 3 years
storageNote1 Refrigerator +4°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.