accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104296-63-1

IUPAC Name: 2-(naphthalen-1-yl)ethane-1-sulfonyl chloride
Synonyms: 2-(naphthalen-1-yl)ethanesulfonyl chloride
Molecular Weight (g/mol): 254.73
Molecular Formula: C12H11ClO2S
InChi Key: IAZQBUVCHCLINF-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)CCC1=CC=CC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)