Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(1-Naphthyl)ethanesulfonyl Chloride 97.0+%, TCI America™
SDP

Catalog No. N05765G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05765G 5 g
N05761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N05765G Supplier TCI America Supplier No. N05765G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 104296-63-1
Molecular Formula C12H11ClO2S
Molecular Weight (g/mol) 254.728
MDL Number MFCD00191560
InChI Key IAZQBUVCHCLINF-UHFFFAOYSA-N
Synonym 2-(1-Naphthyl)ethylsulfonyl Chloride
PubChem CID 21901305
IUPAC Name 2-naphthalen-1-ylethanesulfonyl chloride
SMILES C1=CC=C2C(=C1)C=CC=C2CCS(=O)(=O)Cl

Specifications

Melting Point 50°C
Quantity 5 g
UN Number 3261
Formula Weight 254.73
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(1-Naphthyl)ethanesulfonyl Chloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.