accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104322-63-6

IUPAC Name: (1R,6S,8S)-11,11-dimethyl-5-oxa-3λ⁶-thia-4-azatetracyclo[6.2.1.0¹,⁶.0⁴,⁶]undecane-3,3-dione
Synonyms: (1R,6S,8S)-11,11-dimethyl-5-oxa-3λ⁶-thia-4-azatetracyclo[6.2.1.0¹,⁶.0⁴,⁶]undecane-3,3-dione
Molecular Weight (g/mol): 229.29
Molecular Formula: C10H15NO3S
InChi Key: GBBJBUGPGFNISJ-KAHWVJTKSA-N
SMILES: CC1(C)[C@H]2CC[C@@]11CS(=O)(=O)N3O[C@@]13C2
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (1)
Quantity 
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)