Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-(2R,8aS)-(Camphorylsulfonyl)oxaziridine, 98+%

Catalog No. AC294580010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC294580010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC294580010 Supplier Thermo Scientific Chemicals Supplier No. 294580010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 104322-63-6 | C10H15NO3S | 229.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 104322-63-6
CAS Min % 98.0
CAS Max % 100.0
Melting Point 170.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Molecular Formula C10H15NO3S
Quantity 1 g
MDL Number MFCD00075319
Fieser 16,61; 17,66
Specific Rotation Condition 46.60 (25.00°C c=2,CHCl3)
Specific Rotation 46.6
Synonym 1r---10-camphorsulfonyl oxaziridine
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform
InChI Key GBBJBUGPGFNISJ-SKVWKECBSA-N
SMILES CC1(C2CCC13CS(=O)(=O)N4C3(C2)O4)C
Molecular Weight (g/mol) 229.3
PubChem CID 86278372
Formula Weight 229.3
Percent Purity 98+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (+)-(2R, 8aS)-(Camphorylsulfonyl)oxaziridine, 99+%, (99+% e.e.)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.