CAS RN 104389-32-4
CAS RN 104389-32-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Promotions Available
Sigma Aldrich Polyisoprene, trans
Sigma Aldrich Polyisoprene, trans
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
2
Poly(isoprene), trans, average M.W. 400.000, Thermo Scientific™
CAS: 104389-32-4