CAS RN 10441-57-3
IUPAC Name:
1-methylpyrrolidine-2-thione
Synonyms:
1-methylpyrrolidine-2-thione
Molecular Weight (g/mol):
115.19
Molecular Formula:
C5H9NS
InChi Key:
OQILOJRSIWGQSM-UHFFFAOYSA-N
SMILES:
CN1CCCC1=S
1
–
1
of
1
results
1
1-Methylpyrrolidine-2-thione 97.0+%, TCI America™
CAS: 10441-57-3 Molecular Formula: C5H9NS Molecular Weight (g/mol): 115.19 MDL Number: MFCD00966332 InChI Key: OQILOJRSIWGQSM-UHFFFAOYSA-N PubChem CID: 575447 IUPAC Name: 1-methylpyrrolidine-2-thione SMILES: CN1CCCC1=S