Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylpyrrolidine-2-thione 97.0+%, TCI America™
SDP

Catalog No. M17485G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M17485G Supplier TCI America Supplier No. M17485G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10441-57-3
Molecular Formula C5H9NS
Molecular Weight (g/mol) 115.19
MDL Number MFCD00966332
InChI Key OQILOJRSIWGQSM-UHFFFAOYSA-N
PubChem CID 575447
IUPAC Name 1-methylpyrrolidine-2-thione
SMILES CN1CCCC1=S

Specifications

Melting Point 16°C
Color Yellow
Boiling Point 145°C
Quantity 5 g
Formula Weight 115.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methylpyrrolidine-2-thione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.