accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104504-43-0

OHOOOONH(S)H3CCH3H3C

CAS RN 104504-43-0

IUPAC Name: (2S)-3-(4-benzoylphenyl)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Synonyms: (2S)-3-(4-benzoylphenyl)-2-[(tert-butoxycarbonyl)amino]propanoic acid
Molecular Weight (g/mol): 369.42
Molecular Formula: C21H23NO5
InChi Key: HIQJNYPOWPXYIC-KRWDZBQOSA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(C=C1)C(=O)C1=CC=CC=C1)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)