accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104515-79-9

IUPAC Name: 2-[4,5-bis(ethylsulfanyl)-2H-1,3-dithiol-2-ylidene]-4,5-bis(ethylsulfanyl)-2H-1,3-dithiole
Synonyms: 2-[4,5-bis(ethylsulfanyl)-1,3-dithiol-2-ylidene]-4,5-bis(ethylsulfanyl)-1,3-dithiole
Molecular Weight (g/mol): 444.79
Molecular Formula: C14H20S8
InChi Key: LLDJYFLUQDMPQI-UHFFFAOYSA-N
SMILES: CCSC1=C(SCC)SC(S1)=C1SC(SCC)=C(SCC)S1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results