Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakis(ethylthio)tetrathiafulvalene [Organic Electronic Material], TCI America™
SDP

Catalog No. T1571100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1571100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1571100MG Supplier TCI America Supplier No. T1571100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 104515-79-9
Molecular Formula C14H20S8
Molecular Weight (g/mol) 444.79
MDL Number MFCD00137866
InChI Key LLDJYFLUQDMPQI-UHFFFAOYSA-N
Synonym TET-TTF
PubChem CID 3524833
IUPAC Name 2-[4,5-bis(ethylsulfanyl)-2H-1,3-dithiol-2-ylidene]-4,5-bis(ethylsulfanyl)-2H-1,3-dithiole
SMILES CCSC1=C(SCC)SC(S1)=C1SC(SCC)=C(SCC)S1

Specifications

Melting Point 72°C
Quantity 100 mg
Formula Weight 444.79
Physical Form Crystalline Powder
Chemical Name or Material Tetrakis(ethylthio)tetrathiafulvalene [Organic Electronic Material]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.