CAS RN 10466-56-5
IUPAC Name:
2-amino-3-methylpentanamide hydrochloride
Synonyms:
2-amino-3-methylpentanamide hydrochloride
Molecular Weight (g/mol):
166.65
Molecular Formula:
C6H15ClN2O
InChi Key:
SAOZPTBFWAEJQL-UHFFFAOYNA-N
SMILES:
Cl.CCC(C)C(N)C(N)=O
1
–
1
of
1
results
1
L-Isoleucinamide Hydrochloride 98.0+%, TCI America™
CAS: 10466-56-5 Molecular Formula: C6H15ClN2O Molecular Weight (g/mol): 166.649 MDL Number: MFCD00058476 InChI Key: SAOZPTBFWAEJQL-FHAQVOQBSA-N PubChem CID: 16219521 IUPAC Name: (2S,3S)-2-amino-3-methylpentanamide;hydrochloride SMILES: CCC(C)C(C(=O)N)N.Cl