accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10466-61-2

IUPAC Name: (2S)-2-amino-4-methylpentanamide hydrochloride
Synonyms: leucinamide hydrochloride
Molecular Weight (g/mol): 166.65
Molecular Formula: C6H15ClN2O
InChi Key: VSPSRRBIXFUMOU-XZNNNFJINA-N
SMILES: Cl.CC(C)C[C@H](N)C(N)=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)