Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Adrenosterone

Catalog No. 2849985G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-499-85G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 28-499-85G Supplier Sigma Aldrich Supplier No. 2849985G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Adrenosterone

Specifications

CAS 10466-61-2
Melting Point 254°C to 256°C
Molecular Formula C6H14N2O -+ HCl
Linear Formula (CH3)2CHCH2CH(NH2)CONH2-+HCl
MDL Number MFCD00013012
Quantity 5 g
Synonym 11-Ketoandrostenedione; 4-Androstene-3,11,17-trione; Androst-4-ene-3,11,17-trione; Reichstein's substance G
Molecular Weight (g/mol) 166.65 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.