accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1047-16-1

IUPAC Name: 5,7,12,14-tetrahydro-5,12-diazapentacene-7,14-dione
Synonyms: quinacridone
Molecular Weight (g/mol): 312.33
Molecular Formula: C20H12N2O2
InChi Key: NRCMAYZCPIVABH-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2NC2=CC3=C(NC4=CC=CC=C4C3=O)C=C12
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)