Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinacridone (purified by sublimation) 99.0+%, TCI America™
SDP

Catalog No. Q00831G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q00831G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q00831G Supplier TCI America Supplier No. Q00831G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1047-16-1
Molecular Formula C20H12N2O2
Molecular Weight (g/mol) 312.33
MDL Number MFCD00059956
InChI Key NRCMAYZCPIVABH-UHFFFAOYSA-N
Synonym 5,12-Dihydroquino[2,3-b]acridine-7,14-dione, Pigment Violet 19
PubChem CID 13976
IUPAC Name 5,7,12,14-tetrahydro-5,12-diazapentacene-7,14-dione
SMILES O=C1C2=CC=CC=C2NC2=CC3=C(NC4=CC=CC=C4C3=O)C=C12

Specifications

Melting Point 390°C
Quantity 1 g
Formula Weight 312.33
Percent Purity ≥99.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Quinacridone (purified by sublimation)
EINECSNumber (5)-1167&(5)-3291
RTECSNumber VA8533500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.