accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10472-88-5

IUPAC Name: 2-bromo-4-fluoro-6-nitroaniline
Synonyms: 2-bromo-4-fluoro-6-nitroaniline
Molecular Weight (g/mol): 235.01
Molecular Formula: C6H4BrFN2O2
InChi Key: HCYDUPDSEDHSQB-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=C(F)C=C1[N+]([O-])=O
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)