Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-fluoro-6-nitroaniline, tech. 90%, Thermo Scientific™

Catalog No. AAH6184006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6184006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6184006 Supplier Thermo Scientific Chemicals Supplier No. H6184006

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Bromo-4-fluoro-6-nitroaniline is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10472-88-5
Molecular Formula C6H4BrFN2O2
Molecular Weight (g/mol) 235.01
MDL Number MFCD07779528
InChI Key HCYDUPDSEDHSQB-UHFFFAOYSA-N
Synonym 2-bromo-4-fluoro-6-nitrobenzenamine, 2-bromo-4-fluoro-6-nitrophenylamine, benzenamine, 2-bromo-4-fluoro-6-nitro, 2-amino-3-bromo-5-fluoronitrobenzene, pubchem16316, ksc495m4t, 4-fluoro-2-bromo-6-nitroaniline, 4-fluoro-2-nitro-6-bromoaniline, 6-bromo-4-fluoro-2-nitroaniline, 2-bromo-4-fluoro-6-nitroaniline
PubChem CID 292004
IUPAC Name 2-bromo-4-fluoro-6-nitroaniline
SMILES NC1=C(Br)C=C(F)C=C1[N+]([O-])=O

Specifications

Color Orange
Quantity 5 g
UN Number UN2811
Formula Weight 235.01
Percent Purity 90%
Grade Technical
Physical Form Powder
Chemical Name or Material 2-Bromo-4-fluoro-6-nitroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.