accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH2OH2OH2OH2OH2ONaNaOHBHOOOBOHHOOO

CAS RN 10486-00-7

IUPAC Name: disodium tetrahydroxy-1,2,4,5,3,6-tetraoxadiborinane-3,6-diuide hexahydrate
Synonyms: disodium perborate hexahydrate
Molecular Weight (g/mol): 307.71
Molecular Formula: B2H16Na2O14
InChi Key: MCYPYXIRGFEFGL-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.[Na+].[Na+].O[B-]1(O)OO[B-](O)(O)OO1
Molecular Weight (g/mol) 
  • (7)
  • (5)
  • (3)
Physical Form 
  • (6)
  • (5)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
Grade 
  • (1)
  • (5)
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (5)
  • (1)
  • (5)
  • (2)
  • (7)
  • (7)
  • (5)
  • (3)
  • (6)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (5)
  • (3)
  • (2)
  • (4)