Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium perborate tetrahydrate, MilliporeSigma™

Catalog No. M1065609050 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 kg
Packaging:
Fibre Carton
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1065609050 50 kg Fibre Carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1065609050 Supplier MilliporeSigma Supplier No. 1065609050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity sodium perborate tetrahydrate suitable for a variety of laboratory applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10486-00-7
Molecular Formula BNaO3 · 4H2O
Molecular Weight (g/mol) 153.86

Specifications

Physical Form Powder
Quantity 50 kg
Linear Formula NaBO3 · 4H2O
Solubility Information 23g/L (20°C)
Formula Weight 153.88
Grade Pure
Packaging Fibre Carton
Density 1.73g/cm3; Bulk Density: 800 kg/m3
Chemical Name or Material Sodium Perborate Tetrahydrate, Sodium perborate tetrahydrate
Recommended Storage Store at +5°C to +30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.