accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHOHOOCH3

CAS RN 10487-05-5

IUPAC Name: 3,3-diethoxypropane-1,2-diol
Synonyms: 3,3-diethoxypropane-1,2-diol
Molecular Weight (g/mol): 164.2
Molecular Formula: C7H16O4
InChi Key: BPNZBKXZVJVEJD-UHFFFAOYNA-N
SMILES: CCOC(OCC)C(O)CO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results