accessibility menu, dialog, popup

UserName

Viewing profile as user:
SCH3∗∗n

CAS RN 104934-50-1

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C10H14S)n
InChi Key: —
SMILES: CCCCCCC1=C(-*)SC(-*)=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (1)
  • (6)
Quantity 
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (6)
  • (2)
  • (2)
  • (1)
  • (6)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)