Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P3HT (OPV grade - 91-94% RR)

Catalog No. H66726ME
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H66726-ME 500 mg
H66726-03 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H66726-ME Supplier Thermo Scientific Chemicals Supplier No. H66726ME
Add to Cart

CAS: 104934-50-1 | (C10H14S)n | NaN g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104934-50-1
Molecular Formula (C10H14S)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00217686
InChI Key DUFPJSOXRHVDOV-UHFFFAOYSA-N
Synonym Poly[3-hexylthiophene-2,5-diyl]
IUPAC Name 3-hexyl-2,5-dimethylthiophene
SMILES CCCCCCC1=C(-*)SC(-*)=C1

Specifications

Quantity 500 mg
Physical Form Solid
Chemical Name or Material P3HT
Recommended Storage Air Sensitive. Store under Nitrogen. Ambient temperatures.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.