accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 105-04-4

IUPAC Name: [2-(diethylamino)ethyl](ethyl)amine
Synonyms: [2-(diethylamino)ethyl](ethyl)amine
Molecular Weight (g/mol): 144.26
Molecular Formula: C8H20N2
InChi Key: HDCAZTXEZQWTIJ-UHFFFAOYSA-N
SMILES: CCNCCN(CC)CC
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)