Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N,N'-Triethylethylenediamine 99+%, Thermo Scientific™

Catalog No. AC155610100
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC155610100 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC155610100 Supplier Thermo Scientific Chemicals Supplier No. 155610100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-04-4
Molecular Formula C8H20N2
Molecular Weight (g/mol) 144.26
MDL Number MFCD00009054
InChI Key HDCAZTXEZQWTIJ-UHFFFAOYSA-N
Synonym n,n,n'-triethylethylenediamine, triethylethylenediamine, n,n,n'-triethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n'-triethyl, 2-diethylamino ethyl ethyl amine, 1,2-ethanediamine, n1,n1,n2-triethyl, ethylenediamine, n,n,n'-triethyl, n1,n1,n2-triethyl-1,2-ethanediamine, n,n,n-triethylenediamine, diethyl 2-ethylaminoethyl amine
PubChem CID 66040
IUPAC Name [2-(diethylamino)ethyl](ethyl)amine
SMILES CCNCCN(CC)CC

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Yellow
Density 0.8
Infrared Spectrum Authentic
Refractive Index 1.4289 to 1.4309 (20°C, 589nm)
Linear Formula (C2H5)2NCH2CH2NHC2H5
Quantity 10 mL
Beilstein 04, II, 691
Formula Weight 144.26
Percent Purity ≥99.0%
Physical Form Liquid
Chemical Name or Material N, N, N'-Triethylethylenediamine, 99+%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.