accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHCH

CAS RN 105-31-7

IUPAC Name: hex-1-yn-3-ol
Synonyms: 1-hexyn-3-ol
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: LTFTWJYRQNTCHI-UHFFFAOYNA-N
SMILES: CCCC(O)C#C
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)