Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexyn-3-ol, MP Biomedicals

Catalog No. ICN208539 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN208539 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN208539 Supplier MP Biomedicals, Inc Supplier No. 0520853905

Chemical Identifiers

CAS 105-31-7
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
InChI Key LTFTWJYRQNTCHI-UHFFFAOYSA-N
Synonym 1-hexyn-3-ol, propylethinylcarbinol, 1-hexyne-3-ol, 3-hydroxy-1-hexyne, 1-propylpropargyl alcohol, 1-hexine-3-ol, acmc-2098fm, dsstox_cid_30088, dsstox_gsid_51536, 4-01-00-02234 beilstein handbook reference
PubChem CID 7746
IUPAC Name hex-1-yn-3-ol
SMILES CCCC(C#C)O

Specifications

Quantity 10 g
Formula Weight 98.15
Chemical Name or Material 1-Hexyn-3-Ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.