CAS RN 105-37-3
Ethyl propionate, 99+%
CAS: 105-37-3 Molecular Formula: C5H10O2 Molecular Weight (g/mol): 102.13 MDL Number: MFCD00009308 InChI Key: FKRCODPIKNYEAC-UHFFFAOYSA-N Synonym: ethyl propionate,propanoic acid, ethyl ester,propionic ester,propionic ether,propionic acid, ethyl ester,ethylpropionate,propionate d'ethyle,propanoic acid ethyl ester,ethyl n-propionate,propionic acid ethyl ester PubChem CID: 7749 ChEBI: CHEBI:41330 IUPAC Name: ethyl propanoate SMILES: CCC(=O)OCC
Ethyl propionate
CAS: 105-37-3 Molecular Formula: C5H10O2 Molecular Weight (g/mol): 102.133 MDL Number: MFCD00009308 InChI Key: FKRCODPIKNYEAC-UHFFFAOYSA-N Synonym: ethyl propionate,propanoic acid, ethyl ester,propionic ester,propionic ether,propionic acid, ethyl ester,ethylpropionate,propionate d'ethyle,propanoic acid ethyl ester,ethyl n-propionate,propionic acid ethyl ester PubChem CID: 7749 ChEBI: CHEBI:41330 IUPAC Name: ethyl propanoate SMILES: CCC(=O)OCC
Ethyl propionate, 99%
CAS: 105-37-3 Molecular Formula: C5H10O2 Molecular Weight (g/mol): 102.133 MDL Number: MFCD00009308 InChI Key: FKRCODPIKNYEAC-UHFFFAOYSA-N Synonym: ethyl propionate,propanoic acid, ethyl ester,propionic ester,propionic ether,propionic acid, ethyl ester,ethylpropionate,propionate d'ethyle,propanoic acid ethyl ester,ethyl n-propionate,propionic acid ethyl ester PubChem CID: 7749 ChEBI: CHEBI:41330 IUPAC Name: ethyl propanoate SMILES: CCC(=O)OCC
Ethyl Propionate 99.0+%, TCI America™
CAS: 105-37-3 Molecular Formula: C5H10O2 Molecular Weight (g/mol): 102.133 MDL Number: MFCD00009308 InChI Key: FKRCODPIKNYEAC-UHFFFAOYSA-N Synonym: ethyl propionate,propanoic acid, ethyl ester,propionic ester,propionic ether,propionic acid, ethyl ester,ethylpropionate,propionate d'ethyle,propanoic acid ethyl ester,ethyl n-propionate,propionic acid ethyl ester PubChem CID: 7749 ChEBI: CHEBI:41330 IUPAC Name: ethyl propanoate SMILES: CCC(=O)OCC
Sigma Aldrich Ethyl propionate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
MP Biomedicals, Inc Ethyl Propionate, MP Biomedicals
CAS: 105-37-3 Molecular Formula: C5H10O2 Molecular Weight (g/mol): 102.133 InChI Key: FKRCODPIKNYEAC-UHFFFAOYSA-N Synonym: ethyl propionate,propanoic acid, ethyl ester,propionic ester,propionic ether,propionic acid, ethyl ester,ethylpropionate,propionate d'ethyle,propanoic acid ethyl ester,ethyl n-propionate,propionic acid ethyl ester PubChem CID: 7749 ChEBI: CHEBI:41330 IUPAC Name: ethyl propanoate SMILES: CCC(=O)OCC