Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Propionate, MP Biomedicals

Catalog No. ICN215182 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN215182 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN215182 Supplier MP Biomedicals, Inc Supplier No. 0521518280

Chemical Identifiers

CAS 105-37-3
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
InChI Key FKRCODPIKNYEAC-UHFFFAOYSA-N
Synonym ethyl propionate, propanoic acid, ethyl ester, propionic ester, propionic ether, propionic acid, ethyl ester, ethylpropionate, propionate d'ethyle, propanoic acid ethyl ester, ethyl n-propionate, propionic acid ethyl ester
PubChem CID 7749
ChEBI CHEBI:41330
IUPAC Name ethyl propanoate
SMILES CCC(=O)OCC

Specifications

Quantity 100 g
Formula Weight 102.13
Chemical Name or Material Ethyl Propionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.