accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOCH3

CAS RN 105-50-0

IUPAC Name: 1,5-diethyl 3-oxopentanedioate
Synonyms: 1,5-diethyl 3-oxopentanedioate
Molecular Weight (g/mol): 202.21
Molecular Formula: C9H14O5
InChi Key: ZSANYRMTSBBUCA-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)CC(=O)OCC
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)