Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl 1,3-acetonedicarboxylate, 97%, Thermo Scientific™

Catalog No. AC173162500
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC173162500 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC173162500 Supplier Thermo Scientific Chemicals Supplier No. 173162500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-50-0
Molecular Formula C9H14O5
Molecular Weight (g/mol) 202.21
MDL Number MFCD00009200
InChI Key ZSANYRMTSBBUCA-UHFFFAOYSA-N
Synonym diethyl 1,3-acetonedicarboxylate, diethyl 3-oxoglutarate, 1,5-diethyl 3-oxopentanedioate, pentanedioic acid, 3-oxo-, diethyl ester, ethyl acetonedicarboxylate, ethyl 3-oxoglutarate, diethyl acetone-1,3-dicarboxylate, diethyl acetone dicarboxylate, 3-oxopentanedioic acid diethyl ester, acetonedicarboxylic acid diethyl ester
PubChem CID 66045
IUPAC Name 1,5-diethyl 3-oxopentanedioate
SMILES CCOC(=O)CC(=O)CC(=O)OCC

Specifications

Density 1.112g/mL
Assay Percent Range 97%
Linear Formula C2H5O2CCH2COCH2CO2C2H5
Quantity 250 mL
Beilstein 03, 791
Formula Weight 202.21
Chemical Name or Material Diethyl 1, 3-acetonedicarboxylate, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.