accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 105-57-7

IUPAC Name: 1,1-diethoxyethane
Synonyms: 1,1-diethoxyethane
Molecular Weight (g/mol): 118.18
Molecular Formula: C6H14O2
InChi Key: DHKHKXVYLBGOIT-UHFFFAOYSA-N
SMILES: CCOC(C)OCC
Boiling Point 
  • (2)
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (7)
Percent Purity 
  • (2)
  • (8)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (7)
  • (2)
  • (8)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (5)
  • (3)