accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHOOH

CAS RN 105-59-9

IUPAC Name: 2-[(2-hydroxyethyl)(methyl)amino]ethan-1-ol
Synonyms: N-methyldiethanolamine
Molecular Weight (g/mol): 119.16
Molecular Formula: C5H13NO2
InChi Key: CRVGTESFCCXCTH-UHFFFAOYSA-N
SMILES: CN(CCO)CCO
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)