Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyldiethanolamine, MP Biomedicals

Catalog No. ICN205872 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN205872 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN205872 Supplier MP Biomedicals, Inc Supplier No. 0520587280

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 105-59-9
Molecular Formula C5H13NO2
Molecular Weight (g/mol) 119.16
MDL Number MFCD00002848
InChI Key CRVGTESFCCXCTH-UHFFFAOYSA-N
Synonym n-methyldiethanolamine, methyldiethanolamine, 2,2'-methylimino diethanol, ethanol, 2,2'-methylimino bis, methyl diethanolamine, n-methyliminodiethanol, n-methyl-2,2'-iminodiethanol, n-methylaminodiglycol, 2,2'-methyliminodiethanol, n-methyldiethanolimine
PubChem CID 7767
IUPAC Name 2-[(2-hydroxyethyl)(methyl)amino]ethan-1-ol
SMILES CN(CCO)CCO

Specifications

Quantity 100 g
Formula Weight 119.16
Chemical Name or Material N-Methyldiethanolamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.