accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 105-65-7

IUPAC Name: (propan-2-yloxy)({[(propan-2-yloxy)methanethioyl]disulfanyl})methanethione
Synonyms: thioperoxydicarbonic acid (((HO)C(S))2S2), bis(1
Molecular Weight (g/mol): 270.44
Molecular Formula: C8H14O2S4
InChi Key: ZWWQICJTBOCQLA-UHFFFAOYSA-N
SMILES: CC(C)OC(=S)SSC(=S)OC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results