Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropylxanthogen Disulfide, MP Biomedicals

Catalog No. ICN213035 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN213035 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN213035 Supplier MP Biomedicals, Inc Supplier No. 0521303550

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 105-65-7
Molecular Formula C8H14O2S4
Molecular Weight (g/mol) 270.438
InChI Key ZWWQICJTBOCQLA-UHFFFAOYSA-N
Synonym diproxide, isopropylxanthic disulfide, diproxid, diisopropyl xanthogen disulfide, o,o-diisopropyl dithiobis thioformate, diisopropylxanthogen, diisopropyl dixanthogen, bis isopropylxanthogen, diisopropylxanthogen disulfide, diisopropyl xanthogenate
PubChem CID 66923
IUPAC Name O-propan-2-yl (propan-2-yloxycarbothioyldisulfanyl)methanethioate
SMILES CC(C)OC(=S)SSC(=S)OC(C)C

Specifications

Quantity 100 g
Formula Weight 270.46
Chemical Name or Material Diisopropylxanthogen Disulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.