accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 105-83-9

IUPAC Name: bis(3-aminopropyl)(methyl)amine
Synonyms: bis(3-aminopropyl)(methyl)amine
Molecular Weight (g/mol): 145.25
Molecular Formula: C7H19N3
InChi Key: KMBPCQSCMCEPMU-UHFFFAOYSA-N
SMILES: CN(CCCN)CCCN
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)