accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10503-96-5

H3COCl

CAS RN 10503-96-5

IUPAC Name: 1-(2-chloroethoxy)butane
Synonyms: 1-(2-chloroethoxy)butane
Molecular Weight (g/mol): 136.62
Molecular Formula: C6H13ClO
InChi Key: OJMVRGXLTANFRG-UHFFFAOYSA-N
SMILES: CCCCOCCCl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results