accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10510-77-7

IUPAC Name: 5-ethylphenazin-5-ium ethyl sulfate
Synonyms: 5-ethylphenazin-5-ium ethyl sulfate
Molecular Weight (g/mol): 334.39
Molecular Formula: C16H18N2O4S
InChi Key: VDJKJPMLWJWQIH-UHFFFAOYSA-M
SMILES: CCOS([O-])(=O)=O.CC[N+]1=C2C=CC=CC2=NC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results