Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenazine Ethosulfate, MP Biomedicals

Catalog No. MP021009565 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021009565 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021009565 Supplier MP Biomedicals, Inc Supplier No. 02100956.5

missing translation for 'validMainframeMsg'

  • Phenazine ethosulfate is an intermediate
  • Used to detect nitric oxide reducatase activity in concert with ascorbic acid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10510-77-7
Molecular Formula C16H18N2O4S
Molecular Weight (g/mol) 334.39
MDL Number MFCD00050286
InChI Key VDJKJPMLWJWQIH-UHFFFAOYSA-M
Synonym phenazine ethosulfate, 5-ethylphenazine, n-ethylphenazonium ethosulfate, 5-ethylphenazinium ethyl sulphate, 5-ethylphenazin-5-ium ethyl sulfate, 5-ethylphenazinium ethylsulfate, phenazinium, 5-ethyl-, ethyl sulfate, 5-ethylphenazine, ethyl sulfate salt, ccris 5619, n-ethyldibenzopyrazine ethyl sulfate salt
PubChem CID 82688
IUPAC Name 5-ethylphenazin-5-ium;ethyl sulfate
SMILES CC[N+]1=C2C=CC=CC2=NC3=CC=CC=C31.CCOS(=O)(=O)[O-]

Specifications

Melting Point 173°C to 175°C (literature)
Color Yellow
Linear Formula C16H18N2O4S
Quantity 500 mg
Solubility Information Soluble in water (10mg/mL - clear, yellow solution) or ethanol (50mg/mL - clear, dark yellow to orange solution)
Formula Weight 334.4
Percent Purity ∼95%
Physical Form Powder
Chemical Name or Material PHENAZINE ETHOSULFATE
Recommended Storage -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.