accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 105104-40-3

IUPAC Name: 3-chloro-2-(methoxymethoxy)prop-1-ene
Synonyms: 3-chloro-2-(methoxymethoxy)prop-1-ene
Molecular Weight (g/mol): 136.58
Molecular Formula: C5H9ClO2
InChi Key: JKHPOPCIKQQRPU-UHFFFAOYSA-N
SMILES: COCOC(=C)CCl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results